Invitrogen
Manufacturer Code:PA584464
Catalog # PIPA584464
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human CYTH2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARF exchange factor; ARF nucleotide-binding site opener; ARNO CTS18.1 cytohesin-2 PSCD2 PSCD2L Sec7p-L Sec7p-like; CTC-273B12.8; Cytohesin-2; PH, SEC7 and coiled-coil domain-containing protein 2; pleckstrin homology, Sec7 and coiled-coil domains 2 (cytohesin-2); pleckstrin homology, Sec7 and coiled/coil domains 2 (cytohesin-2); Protein ARNO
Gene Aliases: ARNO; CTS18; CTS18.1; CYTH2; PSCD2; PSCD2L; SEC7L; Sec7p-L; Sec7p-like
UniProt ID: (Human) Q99418
Entrez Gene ID: (Human) 9266
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.