Proteintech
Manufacturer Code:133171AP150UL
Catalog # 50-172-8648
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Cytoglobin Fusion Protein Ag4161 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CYGB cytoglobin HGB Histoglobin STAP; Cytoglobin; HGb; Histoglobin; stellate cell activation associated protein; Stellate cell activation-associated protein
Gene Aliases: 3110001K20Rik; CYGB; HGB; Staap; STAP
UniProt ID: (Human) Q8WWM9, (Mouse) Q9CX80, (Rat) Q921A4
Entrez Gene ID: (Human) 114757, (Mouse) 114886, (Rat) 170520
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.