Novus Biologicals
Manufacturer Code:NBP267020
Catalog # NBP267020
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JM44-23 |
Immunogen | Recombinant Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: Cholesterol 25-hydroxylase; CPD6P450DB1 CYP2D CYP2D7AP CYP2D7BP CYP2D7P2 CYP2D8P2 CYP2DL1 CYPIID6 cytochrome P450 2D6 cytochrome P450 family 2 subfamily D polypeptide 6 cytochrome P450 family 2 subfamily D polypeptide 7 pseudogene 2 cytochrome P450 family 2 subfamily D polypeptide 8 pseudogene 2 cytochrome P450 subfamily II (debrisoquine sparteine etc. -metabolising) cytochrome P450 subfamily IID (debrisoquine sparteine etc. cytochrome P450 subfamily IID (debrisoquine sparteine etc. -metabolising) cytochrome P450 subfamily IID (debrisoquine sparteine etc. -metabolizing) Cytochrome P450-DB1 Debrisoquine 4-hydroxylase EC 1.14.14.1 flavoprotein-linked monooxygenase -metabolizing)-like 1 MGC120389 MGC120390 microsomal monooxygenase P450C2D P450-DB1 polypeptide 6 polypeptide 7 pseudogene 2 polypeptide 8 pseudogene 2 xenobiotic monooxygenase; CYPIID6; Cytochrome P450 2D6; cytochrome P450, family 2, subfamily D, polypeptide 6; cytochrome P450, family 2, subfamily D, polypeptide 7 pseudogene 2; cytochrome P450, family 2, subfamily D, polypeptide 8 pseudogene 2; cytochrome P450, subfamily II (debrisoquine, sparteine, etc., -metabolising), polypeptide 7 pseudogene 2; cytochrome P450, subfamily IID (debrisoquine, sparteine, etc., -metabolising), polypeptide 8 pseudogene 2; cytochrome P450, subfamily IID (debrisoquine, sparteine, etc., -metabolizing), polypeptide 6; cytochrome P450, subfamily IID (debrisoquine, sparteine, etc., -metabolizing)-like 1; Cytochrome P450-DB1; Debrisoquine 4-hydroxylase; flavoprotein-linked monooxygenase; microsomal monooxygenase; nonfunctional cytochrome P450 2D6; xenobiotic monooxygenase
Gene Aliases: CPD6; CYP2D; CYP2D6; CYP2D7AP; CYP2D7BP; CYP2D7P2; CYP2D8P2; CYP2DL1; CYPIID6; P450-DB1; P450C2D; P450DB1
UniProt ID: (Human) P10635
Entrez Gene ID: (Human) 1565
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.