Invitrogen
Manufacturer Code:MA511823
Catalog # PIMA511823
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CTC03 (2B5) |
Immunogen | Equine cytochrome c |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Cytochrome c; Cytochrome c, somatic; cytochrome c-like protein
Gene Aliases: CYC; CYCS; cytc; HCS; THC4
UniProt ID: (Dog) P00011, (Bovine) P62894, (Human) P99999, (Mouse) P62897
Entrez Gene ID: (Dog) 475258, (Bovine) 510767, (Human) 54205, (Horse) 100053958, (Mouse) 13063
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.