Novus Biologicals
Manufacturer Code:NBP2444470.2MG
Catalog # NBP24444702
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 7H8.2C12 + CYCS/1010 |
Immunogen | Synthetic peptides corresponding to amino acid 1-80 81-104 and 66-104 of pigeon cytochrome c (7H8.2C12) Recombinant full-length human CYCS protein (CYCS/1010) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CYCHCS cytochrome c cytochrome c somatic THC4; Cytochrome c
Gene Aliases: CYC; CYCS; HCS; THC4
UniProt ID: (Human) P99999
Entrez Gene ID: (Human) 54205
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.