Proteintech
Manufacturer Code:123651AP150UL
Catalog # 50-172-7928
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Cytochrome b5 Fusion Protein Ag3038 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CYB5 CYB5A Cytochrome b5 MCB5; cytochrome b-5; Cytochrome b5; cytochrome b5 type A (microsomal); MCB5; Microsomal cytochrome b5 type A; type 1 cyt-b5
Gene Aliases: 0610009N12Rik; CYB5; CYB5A; MCB5
UniProt ID: (Human) P00167, (Rat) P00173, (Mouse) P56395
Entrez Gene ID: (Human) 1528, (Rat) 64001, (Mouse) 109672
Molecular Function:
oxidase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.