Invitrogen
Manufacturer Code:PA538907
Catalog # PIPA538907
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human Cytochrome P450 2C8/9/18/19 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: (R)-limonene 6-monooxygenase; (S)-limonene 6-monooxygenase; (S)-limonene 7-monooxygenase; (S)-mephenytoin hydroxylase associated cytochrome P450; Cholesterol 25-hydroxylase; CYPIIC17; CYPIIC18; CYPIIC19; CYPIIC8; CYPIIC9; cytochrome P-450 II C; cytochrome P-450 S-mephenytoin 4-hydroxylase; Cytochrome P-450MP; Cytochrome P450 2C18; Cytochrome P450 2C19; Cytochrome P450 2C8; Cytochrome P450 2C9; Cytochrome P450 2C9 (R)-limonene 6-monooxygenase (S)-limonene 6-monooxygenase (S)-limonene 7-monooxygenase CYPIIC9 Cytochrome P-450MP Cytochrome P450 MP-4 Cytochrome P450 MP-8 Cytochrome P450 PB-1 S-mephenytoin 4-hydroxylase CYP2C9 CYP2C10 Cytochrome P450 2C8 CYPIIC8 Cytochrome P450 IIC2 Cytochrome P450 MP-12 Cytochrome P450 MP-20 Cytochrome P450 form 1 S-mephenytoin 4-hydroxylase CYP2C8 Cytochrome P450 2C18 CYPIIC18 Cytochrome P450-6b/29c CYP2C18 Cytochrome P450 2C19 (R)-limonene 6-monooxygenase (S)-limonene 6-monooxygenase (S)-limonene 7-monooxygenase CYPIIC17 CYPIIC19 Cytochrome P450-11A Cytochrome P450-254C Mephenytoin 4-hydroxylase CYP2C19; Cytochrome P450 form 1; Cytochrome P450 IIC2; Cytochrome P450 MP-12; Cytochrome P450 MP-20; Cytochrome P450 MP-4; Cytochrome P450 MP-8; Cytochrome P450 PB-1; cytochrome P450, family 2, subfamily C, polypeptide 18; cytochrome P450, family 2, subfamily C, polypeptide 19; cytochrome P450, family 2, subfamily C, polypeptide 8; cytochrome P450, family 2, subfamily C, polypeptide 9; cytochrome P450, subfamily IIC (mephenytoin 4-hydroxylase), polypeptide 17; cytochrome P450, subfamily IIC (mephenytoin 4-hydroxylase), polypeptide 18; cytochrome P450, subfamily IIC (mephenytoin 4-hydroxylase), polypeptide 19; cytochrome P450, subfamily IIC (mephenytoin 4-hydroxylase), polypeptide 8; Cytochrome P450-11A; Cytochrome P450-254C; Cytochrome P450-6b/29c; Fenbendazole monooxygenase (4'-hydroxylating); flavoprotein-linked monooxygenase; mephenytoin 4'-hydroxylase; Mephenytoin 4-hydroxylase; microsomal monooxygenase; P450 form 1; S-mephenytoin 4-hydroxylase; unspecific monooxygenase; xenobiotic monooxygenase
Gene Aliases: CPC8; CPC9; CPCI; CPCJ; CYP2C; CYP2C10; CYP2C17; CYP2C18; CYP2C19; CYP2C8; CYP2C9; CYPIIC17; CYPIIC19; CYPIIC8; CYPIIC9; MP-12/MP-20; P450-6B/29C; P450C2C; P450IIC17; P450IIC19; P450IIC9
UniProt ID: (Human) P33260, (Human) P33261, (Human) P10632, (Human) P11712
Entrez Gene ID: (Human) 1562, (Human) 1557, (Human) 1558, (Human) 1559
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.