Novus Biologicals
Manufacturer Code:NBP169423
Catalog # NBP169423
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to CYP4F11(cytochrome P450 family 4 subfamily F polypeptide 11) The peptide sequence was selected from the N terminal of CYP4F11. Peptide sequence FPQPPKQNWFWGHQGLVTPTEEGMKTLTQLVTTYPQGFKLWLGPTFPLLI. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-hydroxy fatty acids omega-hydroxylase CYP4F11; CYPIVF11; CYPIVF11 cytochrome P450 4F11 cytochrome P450 family 4 subfamily F polypeptide 11 cytochrome P450 subfamily IVF polypeptide 11 EC 1.14.14.1; Cytochrome P450 4F11; cytochrome P450, family 4, subfamily F, polypeptide 11; cytochrome P450, subfamily IVF, polypeptide 11; Docosahexaenoic acid omega-hydroxylase; Long-chain fatty acid omega-monooxygenase; Phylloquinone omega-hydroxylase CYP4F11
Gene Aliases: CYP4F11; CYPIVF11
UniProt ID: (Human) Q9HBI6
Entrez Gene ID: (Human) 57834
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.