Novus Biologicals
Manufacturer Code:NBP250212AF647
Catalog # NBP250212AH
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | M25-P2A10 |
Immunogen | Peptide Sequence - KNGIHLRLR (amino acids 499 - 507) |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 20-HETE synthase; 20-HETE synthase CP4Y CYP4A2 cytochrome P450 4A11 cytochrome P450 family 4 subfamily A polypeptide 11 cytochrome P450 subfamily IVA polypeptide 11 cytochrome P-450HK-omega cytochrome P450HL-omega fatty acid omega-hydroxylase lauric acid omega-hydroxylase P450HL-omega; 20-hydroxyeicosatetraenoic acid synthase; alkane-1 monooxygenase; CYP4AII; CYPIVA11; Cytochrome P-450HK-omega; Cytochrome P450 4A11; cytochrome P450, family 4, subfamily A, polypeptide 11; cytochrome P450, subfamily IVA, polypeptide 11; Cytochrome P450HL-omega; Fatty acid omega-hydroxylase; Lauric acid omega-hydroxylase; Long-chain fatty acid omega-monooxygenase; P450HL-omega
Gene Aliases: CP4Y; CYP4A11; CYP4A2; CYP4AII
UniProt ID: (Human) Q02928
Entrez Gene ID: (Human) 1579
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.