Novus Biologicals
Manufacturer Code:NBP1781500.05MG
Catalog # NBP17815005
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Dot blot (DB) | Assay dependent |
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Fruit fly |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide taken form the Spook protein form within the C-terminal region (amino acid 500-543) the peptide was post-synthetically modified and conjugated to carrier protein to achieve desired antigenicity. The Spook peptide sequence was conserved in rat mouse and human protein and also present in other species. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
Protein Aliases: 307a1 CG10594 Cyp307a1 CYP307A1-Dm DmelCG10594 Dmspo Spo spo spook; CG10594-PA; CG10594-PC; CYPCCCVIIA1; Cytochrome P450 307a1; Protein spook; spo-PA; spo-PC
Gene Aliases: 307a1; CG10594; CYP307A1; CYP307A1-Dm; Dmel\CG10594; Dmel_CG10594; Dmspo; Spo
UniProt ID: (Fruit fly) Q9VRM7
Entrez Gene ID: (Fruit fly) 38631
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.