Novus Biologicals
Manufacturer Code:NBP270528APC
Catalog # NB270528APC
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1D7 |
Immunogen | Full length recombinant protein of human Cytochrome P450 2C9 (NP_000762) produced in HEK293T cells. |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: (R)-limonene 6-monooxygenase; (R)-limonene 6-monooxygenase (S)-limonene 6-monooxygenase (S)-limonene 7-monooxygenase CPC9 CYP2C CYP2C10 CYPIIC9 cytochrome P450 2C9 Cytochrome P450 MP-4 Cytochrome P450 MP-8 Cytochrome P450 PB-1 cytochrome P-450 S-mephenytoin 4-hydroxylase cytochrome P450 family 2 subfamily C polypeptide 9 cytochrome P450 subfamily IIC (mephenytoin 4-hydroxylase) polypeptide 9 Cytochrome P-450MP EC 1.14.13.48 EC 1.14.13.49 EC 1.14.13.80 EC 1.14.14.1 flavoprotein-linked monooxygenase MGC149605 MGC88320 microsomal monooxygenase P450IIC9 S-mephenytoin 4-hydroxylase xenobiotic monooxygenase; (S)-limonene 6-monooxygenase; (S)-limonene 7-monooxygenase; Cholesterol 25-hydroxylase; CYPIIC9; cytochrome P-450 S-mephenytoin 4-hydroxylase; Cytochrome P-450MP; Cytochrome P450 2C9; Cytochrome P450 MP-4; Cytochrome P450 MP-8; Cytochrome P450 PB-1; cytochrome P450, family 2, subfamily C, polypeptide 9; flavoprotein-linked monooxygenase; microsomal monooxygenase; S-mephenytoin 4-hydroxylase; xenobiotic monooxygenase
Gene Aliases: CPC9; CYP2C; CYP2C10; CYP2C9; CYPIIC9; P450IIC9
UniProt ID: (Human) P11712
Entrez Gene ID: (Human) 1559
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.