Novus Biologicals
Manufacturer Code:NBP250195AF350
Catalog # NP2019A30
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | T5P3G2&asteriskA3 |
Immunogen | Ovalbumin-conjugated synthetic peptide DSNQNEILPE (C-terminal sequence) |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: CYP26A2 Cytochrome P450 26A2 cytochrome P450 26B1 cytochrome P450 CYP26B1 variant 2 Cytochrome P450 retinoic acid-inactivating 2 cytochrome P450 retinoid metabolizing protein cytochrome P450 family 26 subfamily B polypeptide 1 Cytochrome P450RAI-2 DKFZp686G0638 EC 1.14 MGC129613 P450RAI2 P450RAI-2 Retinoic acid-metabolizing cytochrome; Cytochrome P450 26A2; Cytochrome P450 26B1; cytochrome P450 family 26 subfamily A member 1; Cytochrome P450 retinoic acid-inactivating 2; cytochrome P450 retinoid metabolizing protein; cytochrome P450, family 26, subfamily B, polypeptide 1; cytochrome P450, subfamily XXVIB, polypeptide 1; Cytochrome P450RAI-2; Retinoic acid-metabolizing cytochrome
Gene Aliases: CYP26A2; CYP26B1; P450RAI-2; P450RAI2; RHFCA
UniProt ID: (Human) Q9NR63
Entrez Gene ID: (Human) 56603
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.