Novus Biologicals
Manufacturer Code:NBP245645
Catalog # NBP245645
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4B11 |
Immunogen | Full length human recombinant protein of human COX6C(NP_004365) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Cytochrome c oxidase polypeptide VIc; Cytochrome c oxidase polypeptide VIc cytochrome c oxidase subunit 6C cytochrome c oxidase subunit VIc cytochrome c oxidase subunit VIc preprotein; Cytochrome c oxidase subunit 6C; cytochrome c oxidase subunit VIc preprotein
Gene Aliases: COX6C
UniProt ID: (Human) P09669
Entrez Gene ID: (Human) 1345
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.