Invitrogen
Manufacturer Code:PA579120
Catalog # PIPA579120
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human Cytochrome C. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cytochrome c; Cytochrome c CYCS CYC; Cytochrome C, expressed in somatic tissues; Cytochrome c, somatic
Gene Aliases: CYC; CYCS; CYCSA; HCS; THC4
UniProt ID: (Human) P99999, (Mouse) P62897, (Rat) P62898
Entrez Gene ID: (Human) 54205, (Mouse) 13063, (Rat) 25309
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.