Novus Biologicals
Manufacturer Code:NBP292543100UL
Catalog # NB026956
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human CDO1 (NP_001792.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CDO; CDO CDO-I cysteine dioxygenase type 1 Cysteine dioxygenase type I cysteine dioxygenase type I EC 1.13.11.20; Cysteine dioxygenase type 1; Cysteine dioxygenase type I; cysteine dioxygenase, type I
Gene Aliases: CDO-I; CDO1
UniProt ID: (Human) Q16878
Entrez Gene ID: (Human) 1036
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.