Invitrogen
Manufacturer Code:PA585727
Catalog # PIPA585727
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human Cystatin C. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARMD11 antibody CST3 antibody Cystatin C antibody Cystatin C antibody; bA218C14.4 (cystatin C); cystatin 3; Cystatin-3; Cystatin-C; epididymis secretory protein Li 2; Gamma-trace; Neuroendocrine basic polypeptide; Post-gamma-globulin
Gene Aliases: ARMD11; CST3; HEL-S-2
UniProt ID: (Human) P01034
Entrez Gene ID: (Human) 1471
Molecular Function: cysteine protease inhibitor enzyme modulator protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.