R&D Systems
Manufacturer Code:AF6154SP
Catalog # AF6154SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant rat Cystatin C Met1-Ala140 Accession # P14841 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: ARMD11 bA218C14.4 (cystatin C) CST3 cystatin 3 cystatin C cystatin C (amyloid angiopathy and cerebral hemorrhage) cystatin-3 cystatin-C Gamma-trace MGC117328 Neuroendocrine basic polypeptide Post-gamma-globulin; bA218C14.4 (cystatin C); cystatin 3; Cystatin-3; Cystatin-C; epididymis secretory protein Li 2; Gamma-trace; Neuroendocrine basic polypeptide; Post-gamma-globulin
Gene Aliases: ARMD11; CST3; HEL-S-2
UniProt ID: (Human) P01034
Entrez Gene ID: (Human) 1471
Molecular Function: cysteine protease inhibitor enzyme modulator protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.