R&D Systems
Manufacturer Code:AF4055
Catalog # AF4055
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant mouse Cyr61/CCN1 Asp176-Gly281 Accession # NP_034646 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: CCN family member 1; CCN family member 1 CCN1 CCN1IGF-binding protein 10 Cysteine-rich angiogenic inducer 61 cysteine-rich angiogenic inducer 61 GIG1 GIG1cysteine-rich heparin-binding protein 61 IBP-10 IGFBP-10 IGFBP10cysteine-rich anigogenic inducer 61 Insulin-like growth factor-binding protein 10 protein CYR61 Protein GIG1; Cellular communication network factor 1; Cysteine-rich angiogenic inducer 61; cysteine-rich heparin-binding protein 61; cysteine-rich, angiogenic inducer, 61; cysteine-rich, anigogenic inducer, 61; IBP-10; IGF-binding protein 10; IGFBP-10; Insulin-like growth factor-binding protein 10; Protein CYR61; Protein GIG1
Gene Aliases: CCN1; CYR61; GIG1; IGFBP10
UniProt ID: (Human) O00622
Entrez Gene ID: (Human) 3491
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.