Proteintech
Manufacturer Code:511571AP150UL
Catalog # 50-173-5921
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Cyp4a12a Fusion Protein Ag0509 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cyp4a12 Cyp4a12a; CYPIVA12; CYPIVA8; Cytochrome P450 4A12; Cytochrome P450 4A12A; cytochrome P450, 4a12; cytochrome P450, family 4, subfamily a, polypeptide 12; Cytochrome P450-KP1; Cytochrome P450-PP1; P450-KP1; P450-PP1
Gene Aliases: BC025936; Cyp4a-8; Cyp4a12; Cyp4a12a; Cyp4a8
UniProt ID: (Mouse) Q91WL5, (Rat) P24464
Entrez Gene ID: (Mouse) 277753, (Rat) 266674
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.