Invitrogen
Manufacturer Code:PA570471
Catalog # PIPA570471
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human Cyp2f2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cyp2f2 Cyp2f-2; CYPIIF2; Cytochrome P450 2F2; cytochrome P450, 2f2; cytochrome P450, family 2, subfamily f, polypeptide 2; cytochrome P450, subfamily 2F, polypeptide 1; Cytochrome P450, subfamily IIF, polypeptide 1; Cytochrome P450-NAH-2; Naphthalene dehydrogenase; Naphthalene hydroxylase; P450-NAH-2
Gene Aliases: Cyp2f; Cyp2f-2; Cyp2f1; Cyp2f2; Cyp2f4; CYP4502F4
UniProt ID: (Mouse) P33267, (Rat) O35293
Entrez Gene ID: (Mouse) 13107, (Rat) 54246
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.