Abnova
Manufacturer Code:PAB13532
Catalog # 89-116-727
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 266-289 of mouse Cyp27b1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Do not freeze. |
Sheep polyclonal antibody raised against synthetic peptide of Cyp27b1.
For Research Use Only
Protein Aliases: 1alpha(OH)ase; 25(OH)D 1alpha-hydroxylase; 25-hydroxyvitamin D(3) 1-alpha-hydroxylase; 25-hydroxyvitamin D-1 alpha hydroxylase, mitochondrial; 25-hydroxyvitamin D-1 alpha hydroxylase, mitochondrial precursor (25-OHD-1 alpha-hydroxylase) (25-hydroxyvitamin D3 1-alpha-hydroxylase) (VD3 1A hydroxylase) (P450C1 alpha) (P450VD1-alpha); 25-hydroxyvitamin D3 1alpha-hydroxylase; 25-OHD-1 alpha-hydroxylase; Calcidiol 1-monooxygenase; Cp2b; Cyp1; Cyp27b; Cyp40; Cytochrome p450 27B1; Cytochrome P450 subfamily XXVIIB polypeptide 1; cytochrome P450, 27b1; cytochrome P450, 40 (25-hydroxyvitamin D3 1 alpha-hydroxylase); Cytochrome P450C1 alpha; Cytochrome P450VD1-alpha; MGC91184; P450c1; P450VD1alpha; Pddr; VD3 1A hydroxylase; Vdd1; Vddr; VddrI; Vdr
Gene Aliases: Cp2b; Cyp1; Cyp27b; Cyp27b1; Cyp40; P450c1; Pddr; Vdd1; Vddr; VddrI; Vdr
UniProt ID: (Mouse) O35084
Entrez Gene ID: (Mouse) 13115
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.