Abnova
Manufacturer Code:MAB11116
Catalog # 89-109-308
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3B8C1 |
Immunogen | Recombinant protein corresponding to rat Cyp1a2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against recombinant protein of rat Cyp1a2.
For Research Use Only
Protein Aliases: Cholesterol 25-hydroxylase; CYPD45; CYPIA2; Cytochrome P-448; Cytochrome P-450d; Cytochrome P450 1A2; Cytochrome P450 subfamily I (aromatic compound-inducible) member A2 (Q42 form d); cytochrome P450, 1a2; cytochrome P450, subfamily I (aromatic compound-inducible), member A2 (Q42, form d); Cytochrome P450-D; Hydroperoxy icosatetraenoate dehydratase; P-448; P-450d; P450-D; RATCYPD45
Gene Aliases: Cyp1a-2; Cyp1a2; CYPD45; P-450d; RATCYPD45
UniProt ID: (Rat) P04799
Entrez Gene ID: (Rat) 24297
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.