Abnova
Manufacturer Code:MAB3813
Catalog # 89-111-607
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to rat Cyp1a1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against recombinant Cyp1a1.
For Research Use Only
Protein Aliases: AHH; AHRR; aryl hydrocarbon hydroxylase; CP11; CYP1; CYP1A1; Cyp45c; Cypc45c; CYPIA1; cytochrome P1-450; Cytochrome P450 1A1; Cytochrome P450 form 6; cytochrome P450 subfamily I (aromatic compound-inducible) member A1 (C6 form c) (C6 form c); cytochrome P450, 1a1; cytochrome P450, subfamily I (aromatic compound-inducible), member A1 (C6, form c) (C6, form c); Cytochrome P450-C; Cytochrome P450-P1; Cytochrome P450MT2; Cytochrome P450MT2A; Cytochrome P450MT2B; dioxin-inducible; flavoprotein-linked monooxygenase; Hydroperoxy icosatetraenoate dehydratase; microsomal monooxygenase; P-450MC; P1-450; P450-C; P450DX; P450form6; P450MT2; xenobiotic monooxygenase
Gene Aliases: AHH; AHRR; CP11; CYP1; Cyp1a-1; Cyp1a1; Cyp45c; Cypc45c; P-450MC; P1-450; P450-C; P450DX
UniProt ID: (Rat) P00185
Entrez Gene ID: (Rat) 24296
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.