Novus Biologicals
Manufacturer Code:NBP197946PECY7
Catalog # N197946PEC7
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3C6 |
Immunogen | Recombinant human PPIC (39-197aa) protein purified from E. coli |
Conjugate | PE-Cyanine7 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. Do not freeze. |
Mouse Monoclonal Antibody
Protein Aliases: Cyclophilin C; Cyclophilin C CYPCMGC3673 EC 5.2.1.8 parvulin peptidyl-prolyl cis-trans isomerase C peptidylprolyl isomerase C (cyclophilin C) PPIase C Rotamase C; parvulin; Peptidyl-prolyl cis-trans isomerase C; peptidylprolyl isomerase C (cyclophilin C); PPIase C; Rotamase C
Gene Aliases: CYPC; PPIC
UniProt ID: (Human) P45877
Entrez Gene ID: (Human) 5480
Molecular Function: isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.