Invitrogen
Manufacturer Code:PA569299
Catalog # PIPA569299
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human Cyclophilin C |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cyclophilin C; CYPC PPIase C cyclophilin C parvulin peptidyl-prolyl cis-trans isomerase C rotamase C; parvulin; Peptidyl-prolyl cis-trans isomerase C; peptidylprolyl isomerase C (cyclophilin C); PPIase C; Rotamase C
Gene Aliases: CyP-20c; CYPC; PPIC
UniProt ID: (Bovine) Q08E11, (Human) P45877, (Mouse) P30412
Entrez Gene ID: (Dog) 481480, (Rabbit) 100009379, (Bovine) 535494, (Horse) 100073095, (Human) 5480, (Pig) 100511313, (Guinea pig) 100734754, (Rat) 291463, (Mouse) 19038
Molecular Function: isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.