R&D Systems
Manufacturer Code:MAB5410
Catalog # MAB5410
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 549205 |
Immunogen | <EM>E. coli</EM>-derived recombinant human Cyclophilin B Asp34-Glu216 Accession # P23284 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Cyclophilin B; Cyclophilin B cyclophilin-like protein CYPB CYPBEC 5.2.1.8 CYP-S1 MGC2224 OI9MGC14109 peptidyl-prolyl cis-trans isomerase B peptidylprolyl isomerase B (cyclophilin B) PPIase B PPIB Rotamase B S-cyclophilin SCYLP; cyclophilin-like protein; CYP-S1; epididymis secretory protein Li 39; Peptidyl-prolyl cis-trans isomerase B; peptidylprolyl isomerase B (cyclophilin B); PPIase B; Rotamase B; S-cyclophilin; SCYLP
Gene Aliases: CYP-S1; CYPB; HEL-S-39; OI9; PPIB; SCYLP
UniProt ID: (Human) P23284
Entrez Gene ID: (Human) 5479
Molecular Function:
isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.