Invitrogen
Manufacturer Code:MA529768
Catalog # PIMA529768
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 1 |
Immunogen | Recombit Mouse PPIA protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: Cyclophilin A; Cyclosporin A-binding protein; CyP A Peptidyl-prolyl cis-trans isomerase A EC 5.2.1.8 PPIase Rotamase Cyclosporin A-binding protein; Peptidyl-prolyl cis-trans isomerase A; Peptidyl-prolyl cis-trans isomerase A, N-terminally processed; PPIase A; Rotamase A; SP18
Gene Aliases: 2700098C05; Cphn; CyP-18; CypA; Ppia
UniProt ID: (Mouse) P17742
Entrez Gene ID: (Mouse) 268373
Molecular Function:
isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.