Invitrogen
Manufacturer Code:PA529752
Catalog # PIPA529752
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 165 of Human Cyclophilin A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cyclophilin A; Cyclosporin A-binding protein; CYPA CYPH PPIase A T cell cyclophilin cyclophilin cyclosporin A-binding protein peptidyl-prolyl cis-trans isomerase A rotamase A; epididymis secretory sperm binding protein Li 69p; Peptidyl-prolyl cis-trans isomerase A; Peptidyl-prolyl cis-trans isomerase A, N-terminally processed; peptidylprolyl isomerase A (cyclophilin A); PPIase A; Rotamase A; SP18; T cell cyclophilin
Gene Aliases: 2700098C05; Cphn; CyP-18; CYPA; CYPH; HEL-S-69p; PPIA
UniProt ID: (Human) P62937, (Mouse) P17742
Entrez Gene ID: (Human) 5478, (Mouse) 268373
Molecular Function: isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.