Invitrogen
Manufacturer Code:MA535640
Catalog # PIMA535640
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1260 |
Immunogen | A synthesized peptide derived from human Cyclophilin 40 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 40 kDa peptidyl-prolyl cis-trans isomerase; 40 kDa peptidyl-prolyl cis-trans isomerase 4930564J03Rik Cyclophilin-40 Cyclophilin-related protein Cyp 40 Cyp D CYP40 CYP-40 CYPD CYP-S1 EC 5.2.1.8 I79_011990 MGC33096 Peptidyl-prolyl cis-trans isomerase D PPIase PPIase D PPID Ppidl Ppif rotamase Rotamase D S-cyclophilin SCYLP; 40 kDa peptidyl-prolyl cis-trans isomerase D; cyclophilin 40; Cyclophilin D; Cyclophilin-40; Cyclophilin-related protein; CYP-40; cytoplasmic cyclophilin D; Peptidyl-prolyl cis-trans isomerase D; PPIase D; Rotamase D; testicular tissue protein Li 147
Gene Aliases: 4930564J03Rik; CYP-40; CYP40; CYPD; PPID; Ppidl; Ppif
UniProt ID: (Human) Q08752, (Mouse) Q9CR16, (Rat) Q6DGG0
Entrez Gene ID: (Human) 5481, (Mouse) 67738, (Rat) 361967
Molecular Function:
isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.