Invitrogen
Manufacturer Code:PA518749
Catalog # PIPA518749
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (DENFHYKHDREG) corresponding to the internal amino acids of PPID |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: 40 kDa peptidyl-prolyl cis-trans isomerase; 40 kDa peptidyl-prolyl cis-trans isomerase D; cyclophilin 40; cyclophilin D; Cyclophilin-40; Cyclophilin-related protein; CYP-40; CYP-40 MGC33096 peptidylprolyl isomerase D (cyclophilin D) 40 kDa peptidyl-prolyl cis-trans isomerase D PPIase cyclophilin 40 cyclophilin D cyclophilin-related protein peptidylprolyl isomerase D rotamase; Peptidyl-prolyl cis-trans isomerase D; PPIase D; Rotamase D; testicular tissue protein Li 147
Gene Aliases: CYP-40; CYP40; CYPD; PPID
UniProt ID: (Human) Q08752
Entrez Gene ID: (Human) 5481
Molecular Function: isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.