Invitrogen
Manufacturer Code:PA552163
Catalog # PIPA552163
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human Cyclin T2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cyclin T2a; cyclin T2b; Cyclin-T2; CycT2; CYCT2 SDS-stable vimentin-bound DNA fragment HEF42VIM22 cyclin T2a cyclin T2b cyclin-T2 subunit of positive elongation transcription factor b; SDS-stable vimentin-bound DNA fragment HEF42VIM22; subunit of positive elongation transcription factor b
Gene Aliases: CCNT2; CYCT2
UniProt ID: (Human) O60583
Entrez Gene ID: (Human) 905
Molecular Function: RNA binding protein enzyme modulator kinase activator kinase modulator mRNA processing factor nucleic acid binding transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.