Invitrogen
Manufacturer Code:PA5100709
Catalog # PIPA5100709
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CCNT2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2900041I18Rik C81304 Ccnt2 cyclin T2 cyclin T2a cyclin T2b cyclin-T2 CYCT2 CycT2a CycT2b SDS-stable vimentin-bound DNA fragment HEF42VIM22 subunit of positive elongation transcription factor b; cyclin T2a; cyclin T2b; Cyclin-T2; CycT2; SDS-stable vimentin-bound DNA fragment HEF42VIM22; subunit of positive elongation transcription factor b
Gene Aliases: 2900041I18Rik; C81304; CCNT2; CYCT2; CycT2a; CycT2b
UniProt ID: (Human) O60583, (Mouse) Q7TQK0
Entrez Gene ID: (Human) 905, (Mouse) 72949, (Rat) 304758
Molecular Function: RNA binding protein enzyme modulator kinase activator kinase modulator mRNA processing factor nucleic acid binding transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.