Proteintech
Manufacturer Code:209921AP150UL
Catalog # 50-173-3195
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCNT CCNT1 Cyclin T cyclin T1 CYCT1 HIVE1; CDK9-associated C-type protein; cyclin C-related protein; cyclin-T; Cyclin-T1; CycT1; human immunodeficiency virus type 1 (HIV-1) expression (elevated) 1
Gene Aliases: 2810478G24Rik; AI115585; CCNT; CCNT1; CYCT1; HIVE1
UniProt ID: (Human) O60563, (Mouse) Q9QWV9
Entrez Gene ID: (Human) 904, (Rat) 315291, (Mouse) 12455
Molecular Function: RNA binding protein enzyme modulator kinase activator kinase modulator mRNA processing factor nucleic acid binding transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.