Invitrogen
Manufacturer Code:MA514336
Catalog # PIMA514336
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | HE12 |
Immunogen | Recombit human cyclin E protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: G1/S-specific cyclin-E1; G1/S-specific cyclin-E1 cyclin Es cyclin Et
Gene Aliases: AW538188; CCNE; CCNE1; CycE1; pCCNE1
UniProt ID: (Human) P24864, (Mouse) Q61457
Entrez Gene ID: (Human) 898, (Mouse) 12447
Molecular Function: enzyme modulator kinase activator kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.