R&D Systems
Manufacturer Code:MAB43142100
Catalog # MAB43142100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 328311 |
Immunogen | <em>E.coli</em>-derived recombinant human Cyclin D1 Met1-Ile295 Accession No. P24385 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 °C as supplied. 1 month 2 to 8 °C under sterile conditions after reconstitution. 6 months -20 to -70 °C under sterile condition |
Mouse Monoclonal Antibody
Protein Aliases: B-cell CLL/lymphoma 1; B-cell lymphoma 1 protein; B-cell lymphoma 1 protein BCL-1 BCL-1 oncogene BCL1D11S287E CCND1 cyclin D1 cyclin D1 (PRAD1: parathyroid adenomatosis 1) Cyl-1 G1/S-specific cyclin D1 G1/S-specific cyclin-D1 PRAD1 PRAD1 oncogene PRAD1B-cell CLL/lymphoma 1 U21B31; BCL-1; BCL-1 oncogene; G1/S-specific cyclin-D1; PRAD1 oncogene
Gene Aliases: BCL1; CCND1; D11S287E; PRAD1; U21B31
UniProt ID: (Human) P24385
Entrez Gene ID: (Human) 595
Molecular Function: enzyme modulator kinase activator kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.