Invitrogen
Manufacturer Code:710428
Catalog # PI710428
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Oligoclonal |
Type | Antibody |
Clone | 17HCLC |
Immunogen | Peptide corresponding to amino acids 167180 of human Cyclin D1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Polyclonal Antibody
For Research Use Only
Protein Aliases: B-cell CLL/lymphoma 1; B-cell CLL/lymphoma 1 B-cell lymphoma 1 protein BCL-1 oncogene G1/S-specific cyclin D1 G1/S-specific cyclin-D1 PRAD1 oncogene BCL1 D11S287E PRAD1 U21B31; B-cell lymphoma 1 protein; BCL-1; BCL-1 oncogene; G1/S-specific cyclin-D1; PRAD1 oncogene
Gene Aliases: BCL1; CCND1; D11S287E; PRAD1; U21B31
UniProt ID: (Human) P24385
Entrez Gene ID: (Human) 595
Molecular Function: enzyme modulator kinase activator kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.