Invitrogen
Manufacturer Code:MA1156
Catalog # PIMA1156
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | X29.2 |
Immunogen | Xenopus laevis cyclin B2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C-B2; CCNB2 Cyclin B2 HsT17299 G2/Mitotic-Specific Cyclin-B2; G2/mitotic-specific cyclin B2; G2/mitotic-specific cyclin-B2
Gene Aliases: CCNB2; CycB2; cyclinb2; HsT17299; XELAEV_18020679mg
UniProt ID: (Human) O95067, (Mouse) P30276, (Xenopus) P13351
Entrez Gene ID: (Human) 9133, (Mouse) 12442, (Xenopus) 397743
Molecular Function:
enzyme modulator
kinase activator
kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.