R&D Systems
Manufacturer Code:MAB60001100
Catalog # MAB60001100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 2061D |
Immunogen | <em>E. coli</em>-derived recombinant human Cyclin B1 Met1-Pro91 Accession # P14635 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: CCNB CCNB1 cyclin B1 G2/mitotic-specific cyclin B1 G2/mitotic-specific cyclin-B1; G2/mitotic-specific cyclin B1; G2/mitotic-specific cyclin-B1
Gene Aliases: CCNB; CCNB1
UniProt ID: (Human) P14635
Entrez Gene ID: (Human) 891
Molecular Function:
enzyme modulator
kinase activator
kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.