Invitrogen
Manufacturer Code:PA534682
Catalog # PIPA534682
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment contains a sequence corresponding to a region within amino acids 1 and 190 of Cyclin A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCN1 CCNA cyclin-A cyclin-A2; Cyclin A; Cyclin A2 (Cyclin A); Cyclin-A; Cyclin-A2
Gene Aliases: AA408589; Ccn-1; CCN1; CCNA; CCNA2; Cyca; CycA2
UniProt ID: (Human) P20248, (Mouse) P51943
Entrez Gene ID: (Human) 890, (Rat) 114494, (Mouse) 12428
Molecular Function:
enzyme modulator
kinase activator
kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.