Abnova
Manufacturer Code:PAB9504
Catalog # 89-134-188
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to mouse Cxcl12. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against a recombinant Cxcl12.
For Research Use Only
Protein Aliases: 12-O-tetradecanoylphorbol 13-acetate repressed protein 1; AI174028; C-X-C motif chemokine 12; PBSF; PBSF/SDF-1; Pre-B cell growth-stimulating factor; pre-B-cell growth-stimulating factor; Scyb12; SDF-1; Sdf1; Sdf1a; Sdf1b; Stromal cell-derived factor 1; Thymic lymphoma cell-stimulating factor; TLSF; TLSF-a; TLSF-b; TPAR1
Gene Aliases: Cxcl12; Pbsf; Scyb12; Sdf1; Tlsf; Tpar1
UniProt ID: (Mouse) P40224
Entrez Gene ID: (Mouse) 20315
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.