Abnova
Manufacturer Code:PAB8798
Catalog # 89-133-943
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to rat Cxcl1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against a recombinant Cxcl1.
For Research Use Only
Protein Aliases: C-X-C motif chemokine 1; chemokine (C-X-C motif) ligand 1 (melanoma growth stimulating activity, alpha); CINC-1; Cytokine-induced neutrophil chemoattractant 1; gro; Gro1; Growth-regulated alpha protein; Platelet-derived growth factor-inducible protein KC
Gene Aliases: CINC-1; Cinc1; Cxcl1; Gro; Gro1; Scyb1
UniProt ID: (Rat) P14095
Entrez Gene ID: (Rat) 81503
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.