Abnova
Manufacturer Code:PAB17974
Catalog # 89-119-587
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to the residues surrounding amino acid 78 of mouse Cxcl1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Cxcl1.
For Research Use Only
Protein Aliases: alpha-chemokine; C-X-C motif chemokine 1; Fsp; gro; Gro1; GRO1 oncogene; Growth-regulated alpha protein; Hematopoietic synergistic factor; HSF; KC; KC(5-72); KC-T; KC/GR)-alpha; KC/GRO-alpha; Mgsa; N51; Platelet-derived growth factor-inducible protein KC; Scyb1; Secretory protein N51
Gene Aliases: Cxcl1; Fsp; Gro; Gro1; KC; Mgsa; N51; Scyb1
UniProt ID: (Mouse) P12850
Entrez Gene ID: (Mouse) 14825
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.