Abnova
Manufacturer Code:PAB19482
Catalog # 89-121-008
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids at N-terminus of mouse Cx3cl1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C on dry atmosphere. After reconstitution with 200&muL of deionized water and concentration will be 500&mu/mL,store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Cx3cl1.
For Research Use Only
Protein Aliases: AB030188; ABCD-3; AI848747; C-X3-C motif chemokine 1; CX3C; CX3C membrane-anchored chemokine; Cxc3; D8Bwg0439e; Fractalkine; Neurotactin; Processed fractalkine; Scyd1; small inducible cytokine subfamily D, 1; Small-inducible cytokine D1
Gene Aliases: AB030188; ABCD-3; AI848747; CX3C; Cx3cl1; Cxc3; D8Bwg0439e; Fkn; Scyd1
UniProt ID: (Mouse) O35188
Entrez Gene ID: (Mouse) 20312
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.