Invitrogen
Manufacturer Code:PA595767
Catalog # PIPA595767
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human CUL3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CUL-3; Cullin-3
Gene Aliases: AI467304; AW146203; CUL-3; CUL3; KIAA0617; mKIAA0617; PHA2E
UniProt ID: (Human) B8ZZC3, (Mouse) Q9JLV5, (Rat) B5DF89
Entrez Gene ID: (Human) 8452, (Mouse) 26554, (Rat) 301555
Molecular Function: ligase ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.