Abnova
Manufacturer Code:PAB8637
Catalog # 89-133-812
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues surrounding amino acids 223 of rat Ctsl1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Ctsl1.
For Research Use Only
Protein Aliases: Cathepsin L; Cathepsin L heavy chain; Cathepsin L light chain; Cathepsin L1; CATHL; CP-2; Ctsl; Cyclic protein 2; Major excreted protein; MEP; Procathepsin L
Gene Aliases: CATHL; CatL; Ctsl; Ctsl1
UniProt ID: (Rat) P07154
Entrez Gene ID: (Rat) 25697
Molecular Function: cysteine protease hydrolase protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.