Abnova
Manufacturer Code:MAB14496
Catalog # 89-034-579
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 25a |
Immunogen | Native purified mouse Ctnnd1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AA409437; AU019353; Cadherin-associated Src substrate; CAS; Catenin delta-1; catenin src; Catns; Ctnnd; mKIAA0384; P120; p120 catenin; p120(cas); p120(ctn); p120-catenin
Gene Aliases: AA409437; AU019353; Catns; Ctnnd; Ctnnd1; Kiaa0384; mKIAA0384; P120
UniProt ID: (Mouse) P30999
Entrez Gene ID: (Mouse) 12388
Molecular Function:
cytoskeletal protein
intermediate filament
intermediate filament binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.