Novus Biologicals
Manufacturer Code:NBP141205FR
Catalog # NBP141205FR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1900000000 |
Immunogen | Human CtIP |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CTBP-interacting protein; CtBP-interacting protein CtIP DNA endonuclease RBBP8 EC 3.1 RBBP-8 retinoblastoma binding protein 8 Retinoblastoma-binding protein 8 Retinoblastoma-interacting protein and myosin-like RIMSAE2 Sporulation in the absence of SPO11 protein 2 homolog; CtIP; DNA endonuclease RBBP8; RBBP-8; retinoblastoma binding protein 8; Retinoblastoma-binding protein 8; Retinoblastoma-interacting protein and myosin-like; RIM; SAE2; Sporulation in the absence of SPO11 protein 2 homolog
Gene Aliases: COM1; CTIP; JWDS; RBBP8; RIM; SAE2; SCKL2
UniProt ID: (Human) Q99708
Entrez Gene ID: (Human) 5932
Molecular Function: transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.