Invitrogen
Manufacturer Code:PA520963
Catalog # PIPA520963
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 14 amino acid peptide near the center of human RBBP8. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CTBP-interacting protein; CtIP; CTIP RIM SAE2 CTBP-interacting protein DNA endonuclease RBBP8 RBBP-8 sporulation in the absence of SPO11 protein 2 homolog; DNA endonuclease RBBP8; RBBP-8; retinoblastoma binding protein 8; Retinoblastoma-binding protein 8; Retinoblastoma-interacting protein and myosin-like; RIM; SAE2; Sporulation in the absence of SPO11 protein 2 homolog
Gene Aliases: 9930104E21Rik; COM1; CTIP; JWDS; RBBP-8; RBBP8; RGD1308872; RIM; SAE2; SCKL2
UniProt ID: (Human) Q99708, (Rat) B1WC58, (Mouse) Q80YR6
Entrez Gene ID: (Human) 5932, (Rat) 291787, (Mouse) 225182
Molecular Function:
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.