Invitrogen
Manufacturer Code:PA579086
Catalog # PIPA579086
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived human CTBP2 recombit protein (Position: H321-Q445). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-terminal-binding protein 2; C-terminal-binding protein 2 CtBP2 CTBP2; CtBP2; gene trap ROSA b-geo 6; ribeye
Gene Aliases: AA407280; CTBP2; D7Ertd45e; Gtrgeo6; Ribeye
UniProt ID: (Human) P56545, (Rat) Q9EQH5, (Mouse) P56546
Entrez Gene ID: (Human) 1488, (Rat) 81717, (Mouse) 13017
Molecular Function:
dehydrogenase
oxidoreductase
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.