Invitrogen
Manufacturer Code:PA110028
Catalog # PIPA110028
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Bacterially expressed full-length human CtBP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BARS MGC104684 C-terminal-binding protein 1 brefeldin A-ribosylated substrate; brefeldin A-ribosylated substrate; C-terminal-binding protein 1; CtBP1
Gene Aliases: BARS; CTBP; CTBP1; CtBP1-L; CtBP1-S; CtBP3/BARS; D4S115h; D5H4S115; D5H4S115E
UniProt ID: (Human) Q13363, (Mouse) O88712
Entrez Gene ID: (Human) 1487, (Mouse) 13016
Molecular Function: dehydrogenase oxidoreductase transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.